Learn More
4-Carboxynaphthalene-1-boronic acid, 95%, Thermo Scientific Chemicals
$477.05 - $1381.38
Chemical Identifiers
CAS | 332398-57-9 |
---|---|
Molecular Formula | C11H9BO4 |
Molecular Weight (g/mol) | 216.00 |
MDL Number | MFCD06203538 |
InChI Key | SRYYKYNBMCQCAB-UHFFFAOYSA-N |
Synonym | 4-carboxynaphthalene-1-boronic acid, 4-borono-1-naphthoic acid, 4-dihydroxyboranyl naphthalene-1-carboxylic acid, acmc-209hzn, 4-carboxy-1-naphthylboronic acid, 4-carboxynaphthalen-1-ylboronic acid |
PubChem CID | 22711863 |
IUPAC Name | 4-borononaphthalene-1-carboxylic acid |
SMILES | OB(O)C1=C2C=CC=CC2=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH56542MD
|
Thermo Scientific Chemicals
H56542MD |
250 mg |
Each for $477.05
|
|
|||||
AAH5654203
|
Thermo Scientific Chemicals
H5654203 |
1 g |
Each for $1,381.38
|
|
|||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
332398-57-9 | |
216.00 | |
SRYYKYNBMCQCAB-UHFFFAOYSA-N | |
22711863 | |
OB(O)C1=C2C=CC=CC2=C(C=C1)C(O)=O |
C11H9BO4 | |
MFCD06203538 | |
4-carboxynaphthalene-1-boronic acid, 4-borono-1-naphthoic acid, 4-dihydroxyboranyl naphthalene-1-carboxylic acid, acmc-209hzn, 4-carboxy-1-naphthylboronic acid, 4-carboxynaphthalen-1-ylboronic acid | |
4-borononaphthalene-1-carboxylic acid |
Specifications
332398-57-9 | |
MFCD06203538 | |
4-carboxynaphthalene-1-boronic acid, 4-borono-1-naphthoic acid, 4-dihydroxyboranyl naphthalene-1-carboxylic acid, acmc-209hzn, 4-carboxy-1-naphthylboronic acid, 4-carboxynaphthalen-1-ylboronic acid | |
OB(O)C1=C2C=CC=CC2=C(C=C1)C(O)=O | |
216.00 | |
216 | |
4-Carboxynaphthalene-1-boronic acid |
C11H9BO4 | |
250 mg | |
SRYYKYNBMCQCAB-UHFFFAOYSA-N | |
4-borononaphthalene-1-carboxylic acid | |
22711863 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only