missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Carboxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 344400050

 View more versions of this product

Catalog No. AC344400050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Carboxyphenylboronic acid
96.0
238.0°C to 240.0°C
Authentic
Glass bottle
HOOCC6H4B(OH)2
5 g
Solubility in water: 25g/L.
OB(O)C1=CC=C(C=C1)C(O)=O
165.94
165.94
Powder
14047-29-1
100.0
White to Beige
97%
C7H7BO4
MFCD00151801
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
SIAVMDKGVRXFAX-UHFFFAOYSA-N
4-boronobenzoic acid
312183
97%

Chemical Identifiers

14047-29-1
165.94
SIAVMDKGVRXFAX-UHFFFAOYSA-N
312183
OB(O)C1=CC=C(C=C1)C(O)=O
C7H7BO4
MFCD00151801
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
4-boronobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only