missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-1,2-difluorobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1292422

 View more versions of this product

Catalog No. AAA1292422


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-1,2-difluorobenzene
1.35
32°C (89°F)
C6H3ClF2
MFCD00042572
UN1993
1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
C1=CC(=C(C=C1Cl)F)F
148.537
148.54
696-02-6
128°C to 130°C
Aromatic
1.4745
100 g
2081081
OPQMRQYYRSTBME-UHFFFAOYSA-N
4-chloro-1,2-difluorobenzene
136519
98%

Chemical Identifiers

696-02-6
148.537
OPQMRQYYRSTBME-UHFFFAOYSA-N
136519
C1=CC(=C(C=C1Cl)F)F
C6H3ClF2
MFCD00042572
1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
4-chloro-1,2-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only