missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-1-butanol, 85%, balance THF and HCl, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 356310100

 View more versions of this product

Catalog No. AC356310100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-1-butanol
928-51-8
100.0
1.0890g/mL
36°C
80% min. (GC)
C4H9ClO
Cl(CH2)4OH
10 g
1.089
HXHGULXINZUGJX-UHFFFAOYSA-N
4-chlorobutan-1-ol
13569
108.57
Liquid
Balance THF and HCl
80.0
Colorless to Yellow
58.0°C to 60.0°C (3.0 mmHg)
Authentic
Glass bottle
1.4520 to 1.4560
MFCD00002967
01, II, 398
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
OCCCCCl
108.57
CHEBI:87310
85%

Chemical Identifiers

928-51-8
108.57
HXHGULXINZUGJX-UHFFFAOYSA-N
13569
4-chlorobutan-1-ol
C4H9ClO
MFCD00002967
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
CHEBI:87310
OCCCCCl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be corrosive to metals.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Absorb spillage to prevent material damage.

GHS Signal Word: Warning

EINECSNumber : 213-175-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only