Learn More
4-Chloro-1-butanol, 85%, balance THF and HCl, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 356311000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-1-butanol | |
928-51-8 | |
100.0 | |
1.0890g/mL | |
36°C | |
80% min. (GC) | |
C4H9ClO | |
Cl(CH2)4OH | |
100 g | |
1.089 | |
HXHGULXINZUGJX-UHFFFAOYSA-N | |
4-chlorobutan-1-ol | |
13569 | |
108.57 | |
Liquid |
Balance THF and HCl | |
80.0 | |
Colorless to Yellow | |
58.0°C to 60.0°C (3.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4520 to 1.4560 | |
MFCD00002967 | |
01, II, 398 | |
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol | |
OCCCCCl | |
108.57 | |
CHEBI:87310 | |
85% |
Chemical Identifiers
928-51-8 | |
108.57 | |
HXHGULXINZUGJX-UHFFFAOYSA-N | |
13569 | |
4-chlorobutan-1-ol |
C4H9ClO | |
MFCD00002967 | |
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol | |
CHEBI:87310 | |
OCCCCCl |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be corrosive to metals.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Absorb spillage to prevent material damage.
GHS Signal Word: Warning
EINECSNumber : 213-175-5
RUO – Research Use Only