Learn More
4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 460160050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chloro-1-butene | |
| 0.9210g/mL | |
| Authentic | |
| C4H7Cl | |
| 0.921 | |
| WKEVRZCQFQDCIR-UHFFFAOYSA-N | |
| 4-chlorobut-1-ene | |
| 13565 | |
| 98% |
| 927-73-1 | |
| −8°C | |
| 97.5% min. (GC) | |
| 5 g | |
| 4-chloro-1-butene, 1-butene, 4-chloro, 1-but-3-enyl chloride, ksc487m9b | |
| C=CCCCl | |
| 90.55 | |
| 90.55 |
Chemical Identifiers
| 927-73-1 | |
| 90.55 | |
| 4-chloro-1-butene, 1-butene, 4-chloro, 1-but-3-enyl chloride, ksc487m9b | |
| 4-chlorobut-1-ene |
| C4H7Cl | |
| WKEVRZCQFQDCIR-UHFFFAOYSA-N | |
| 13565 | |
| C=CCCCl |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Store in a well-ventilated place. Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 213-160-3
RUO – Research Use Only