Learn More
4'-Chloro-2,2':6',2″-terpyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L14728MD
Description
4'-Chloro-2,2':6',2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture double helicates, dendrimers, micelles and metallo-supramolecular polymers. It is used to synthesize the mono- and bis-terpyridines. It is actively involved in Williamson type ether reactions alpha,μ-bishydroxy-functionalized poly(propylene oxide).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Chloro-2,2′:6′,2′′-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture double helicates, dendrimers, micelles and metallo-supramolecular polymers. It is used to synthesize the mono- and bis-terpyridines. It is actively involved in Williamson type ether reactions alpha,µ-bishydroxy-functionalized poly(propylene oxide).
Solubility
Slightly soluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
4'-Chloro-2,2':6',2''-terpyridine | |
148°C to 152°C | |
MFCD00191930 | |
3613386 | |
Slightly soluble in water. | |
ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 | |
267.72 | |
267.72 |
128143-89-5 | |
C15H10ClN3 | |
250 mg | |
4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine | |
AHEMFMCEBIJRMU-UHFFFAOYSA-N | |
4-chloro-2,6-dipyridin-2-ylpyridine | |
667748 | |
98% |
Chemical Identifiers
128143-89-5 | |
267.72 | |
AHEMFMCEBIJRMU-UHFFFAOYSA-N | |
667748 |
C15H10ClN3 | |
MFCD00191930 | |
4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine | |
ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H315-H318-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only