Learn More
4-Chloro-2,6-dimethylaniline, 96%, Thermo Scientific Chemicals
$105.03 - $337.44
Chemical Identifiers
CAS | 24596-18-7 |
---|---|
Molecular Formula | C8H10ClN |
Molecular Weight (g/mol) | 155.625 |
MDL Number | MFCD01076576 |
InChI Key | JCLZLZKUISPXDC-UHFFFAOYSA-N |
Synonym | 4-chloro-2,6-dimethyl-phenylamine, 4-chloro-2,6-dimethylbenzenamine, 2,6-dimethyl-4-chloroaniline, intermediates-zcf02661, acmc-20a53k, 4-chlor-2,6-dimethylanilin, pharmabridge p-1739, 4-chloro-2,6-dimethyl-aniline, 4-chloro-2,6-xylidine, 4-chloro-2,6-dimethylphenylamine |
PubChem CID | 5073362 |
IUPAC Name | 4-chloro-2,6-dimethylaniline |
SMILES | CC1=CC(=CC(=C1N)C)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2199606
|
Thermo Scientific Chemicals
B2199606 |
5 g |
Each for $105.03
|
|
|||||
AAB2199614
|
Thermo Scientific Chemicals
B2199614 |
25 g |
Each for $337.44
|
|
|||||
Description
4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
24596-18-7 | |
155.625 | |
JCLZLZKUISPXDC-UHFFFAOYSA-N | |
5073362 | |
CC1=CC(=CC(=C1N)C)Cl |
C8H10ClN | |
MFCD01076576 | |
4-chloro-2,6-dimethyl-phenylamine, 4-chloro-2,6-dimethylbenzenamine, 2,6-dimethyl-4-chloroaniline, intermediates-zcf02661, acmc-20a53k, 4-chlor-2,6-dimethylanilin, pharmabridge p-1739, 4-chloro-2,6-dimethyl-aniline, 4-chloro-2,6-xylidine, 4-chloro-2,6-dimethylphenylamine | |
4-chloro-2,6-dimethylaniline |
Specifications
45°C to 50°C | |
5 g | |
MFCD01076576 | |
Slightly soluble in water. | |
CC1=CC(=CC(=C1N)C)Cl | |
155.625 | |
155.63 | |
4-Chloro-2,6-dimethylaniline |
24596-18-7 | |
C8H10ClN | |
4-chloro-2,6-dimethyl-phenylamine, 4-chloro-2,6-dimethylbenzenamine, 2,6-dimethyl-4-chloroaniline, intermediates-zcf02661, acmc-20a53k, 4-chlor-2,6-dimethylanilin, pharmabridge p-1739, 4-chloro-2,6-dimethyl-aniline, 4-chloro-2,6-xylidine, 4-chloro-2,6-dimethylphenylamine | |
JCLZLZKUISPXDC-UHFFFAOYSA-N | |
4-chloro-2,6-dimethylaniline | |
5073362 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only