missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluorotoluene 98.0+%, TCI America™
$46.60 - $281.48
Chemical Identifiers
| CAS | 452-75-5 |
|---|---|
| Molecular Formula | C7H6ClF |
| Molecular Weight (g/mol) | 144.57 |
| MDL Number | MFCD00000571 |
| InChI Key | MKFCYQTVSDCXAQ-UHFFFAOYSA-N |
| Synonym | 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene |
| PubChem CID | 520630 |
| IUPAC Name | 4-chloro-2-fluoro-1-methylbenzene |
| SMILES | CC1=CC=C(Cl)C=C1F |
Chemical Identifiers
| 452-75-5 | |
| 144.57 | |
| MKFCYQTVSDCXAQ-UHFFFAOYSA-N | |
| 520630 | |
| CC1=CC=C(Cl)C=C1F |
| C7H6ClF | |
| MFCD00000571 | |
| 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene | |
| 4-chloro-2-fluoro-1-methylbenzene |
Specifications
| 452-75-5 | |
| C7H6ClF | |
| 25 g | |
| 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene | |
| CC1=CC=C(Cl)C=C1F | |
| 144.57 | |
| 144.57 | |
| Liquid |
| Yellow | |
| MFCD00000571 | |
| 1993 | |
| MKFCYQTVSDCXAQ-UHFFFAOYSA-N | |
| 4-chloro-2-fluoro-1-methylbenzene | |
| 520630 | |
| ≥98.0% (GC) | |
| 4-Chloro-2-fluorotoluene |
Safety and Handling
TSCA : No