missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals

$180.42 - $642.81

Chemical Identifiers

CAS 63069-48-7
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD01863737
InChI Key FLEJOBRWKBPUOX-UHFFFAOYSA-N
Synonym 2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
PubChem CID 2757615
IUPAC Name 4-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)I)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2545706
View Documents
Thermo Scientific Chemicals
B2545706
5 g
Each for $180.42
 
AAB2545714
View Documents
Thermo Scientific Chemicals
B2545714
25 g
Each for $642.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63069-48-7
253.467
FLEJOBRWKBPUOX-UHFFFAOYSA-N
2757615
C1=CC(=C(C=C1Cl)I)N
C6H5ClIN
MFCD01863737
2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
4-chloro-2-iodoaniline

Specifications

63069-48-7
>110(230°F)
MFCD01863737
UN2811
2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
C1=CC(=C(C=C1Cl)I)N
253.467
253.47
4-Chloro-2-iodoaniline
41°C to 44°C
C6H5ClIN
5 g
Light sensitive
FLEJOBRWKBPUOX-UHFFFAOYSA-N
4-chloro-2-iodoaniline
2757615
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 263-839-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only