missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-methyl-6-nitroaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2225303

 View more versions of this product

Catalog No. AAB2225303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-2-methyl-6-nitroaniline
121°C to 123°C
MFCD01320687
4-chloro-2-methyl-6-nitrophenylamine, acmc-1b9ic, 2-amino-5-chloro-3-nitrotoluene, 2-methyl-4-chloro-6-nitroaniline, 4-chloro-2-methyl-6-nitrobenzenamine, 2-amino-5-chloro-3-methylnitrobenzene, benzenamine,4-chloro-2-methyl-6-nitro, benzenamine, 4-chloro-2-methyl-6-nitro
CC1=CC(Cl)=CC(=C1N)[N+]([O-])=O
186.60
186.6
62790-50-5
C7H7ClN2O2
1 g
QDSCDFKGUAONPC-UHFFFAOYSA-N
4-chloro-2-methyl-6-nitroaniline
182355
98%

Chemical Identifiers

62790-50-5
186.60
QDSCDFKGUAONPC-UHFFFAOYSA-N
4-chloro-2-methyl-6-nitroaniline
C7H7ClN2O2
MFCD01320687
182355
CC1=CC(Cl)=CC(=C1N)[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only