Learn More
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 109061000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-3, 5-dimethylphenol | |
88-04-0 | |
100.0 | |
White to Beige | |
132°C | |
98.5% min. (GC) | |
C8H9ClO | |
MFCD00002324 | |
06, II, 463 | |
chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
4-chloro-3,5-dimethylphenol | |
2723 | |
156.61 | |
Crystalline Powder or Crystals |
99% | |
98.5 | |
113.0°C to 116.0°C | |
246.0°C | |
Authentic | |
Plastic bottle | |
ClC6H2(CH3)2OH | |
100 g | |
15, 2182 | |
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,ether and benzene | |
CC1=CC(=CC(=C1Cl)C)O | |
156.61 | |
CHEBI:34393 | |
99% |
Chemical Identifiers
88-04-0 | |
156.61 | |
OSDLLIBGSJNGJE-UHFFFAOYSA-N | |
2723 | |
4-chloro-3,5-dimethylphenol |
C8H9ClO | |
MFCD00002324 | |
chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson | |
CHEBI:34393 | |
CC1=CC(=CC(=C1Cl)C)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 201-793-8
RTECSNumber : ZE6850000
TSCA : TSCA
RUO – Research Use Only