missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylbenzeneboronic acid, 98%

CAS: 161950-10-3 | C7H8BClO2 | 170.40 g/mol

$137.16 - $538.42

Chemical Identifiers

CAS 161950-10-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.40
MDL Number MFCD02683104
InChI Key UZDPQDBLCJDUAX-UHFFFAOYSA-N
Synonym 4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
PubChem CID 3786632
IUPAC Name (4-chloro-3-methylphenyl)boronic acid
SMILES CC1=C(Cl)C=CC(=C1)B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2317903
View Documents
Thermo Scientific Chemicals
B2317903
1 g N/A
Only null left
 
AAB2317906
View Documents
Thermo Scientific Chemicals
B2317906
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

161950-10-3
170.40
UZDPQDBLCJDUAX-UHFFFAOYSA-N
3786632
CC1=C(Cl)C=CC(=C1)B(O)O
C7H8BClO2
MFCD02683104
4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
(4-chloro-3-methylphenyl)boronic acid

Specifications

238°C to 242°C
1 g
MFCD02683104
UZDPQDBLCJDUAX-UHFFFAOYSA-N
(4-chloro-3-methylphenyl)boronic acid
3786632
98%
161950-10-3
C7H8BClO2
4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
CC1=C(Cl)C=CC(=C1)B(O)O
170.40
170.4
4-Chloro-3-methylbenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only