missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 109481000

 View more versions of this product

Catalog No. AC109481000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-3-methylphenol
59-50-7
100.0
White
118°C
99% min. (GC)
C7H7ClO
MFCD00002323
06, 381
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
CFKMVGJGLGKFKI-UHFFFAOYSA-N
4-chloro-3-methylphenol
1732
142.58
Tablets
99+%
99.0
63.0°C to 65.0°C
242°C
Authentic
Plastic bottle
ClC6H3(CH3)OH
100 g
15, 2133
Solubility in water: 4g/L (20 c). Other solubilities: soluble in alkalies,soluble in most common organic solvents
CC1=C(C=CC(=C1)O)Cl
142.58
CHEBI:34395
99+%

Chemical Identifiers

59-50-7
142.58
CFKMVGJGLGKFKI-UHFFFAOYSA-N
1732
4-chloro-3-methylphenol
C7H7ClO
MFCD00002323
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
CHEBI:34395
CC1=C(C=CC(=C1)O)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effect

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

EINECSNumber : 200-431-6

RTECSNumber : GO7100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only