Learn More
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 109485000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-3-methylphenol | |
59-50-7 | |
100.0 | |
White | |
118°C | |
99% min. (GC) | |
C7H7ClO | |
MFCD00002323 | |
06, 381 | |
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan | |
CFKMVGJGLGKFKI-UHFFFAOYSA-N | |
4-chloro-3-methylphenol | |
1732 | |
142.58 | |
Tablets |
99+% | |
99.0 | |
63.0°C to 65.0°C | |
242°C | |
Authentic | |
Plastic bottle | |
ClC6H3(CH3)OH | |
500 g | |
15, 2133 | |
Solubility in water: 4g/L (20 c). Other solubilities: soluble in alkalies,soluble in most common organic solvents | |
CC1=C(C=CC(=C1)O)Cl | |
142.58 | |
CHEBI:34395 | |
99+% |
Chemical Identifiers
59-50-7 | |
142.58 | |
CFKMVGJGLGKFKI-UHFFFAOYSA-N | |
1732 | |
4-chloro-3-methylphenol |
C7H7ClO | |
MFCD00002323 | |
chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan | |
CHEBI:34395 | |
CC1=C(C=CC(=C1)O)Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effect
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
EINECSNumber : 200-431-6
RTECSNumber : GO7100000
TSCA : TSCA
RUO – Research Use Only