Learn More
4-Chloro-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
$184.86 - $184.86
Chemical Identifiers
CAS | 96-99-1 |
---|---|
Molecular Formula | C7H4ClNO4 |
Molecular Weight (g/mol) | 201.562 |
MDL Number | MFCD00007079 |
InChI Key | DFXQXFGFOLXAPO-UHFFFAOYSA-N |
Synonym | 4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene |
PubChem CID | 7320 |
SMILES | C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1280736
|
Thermo Scientific Chemicals
A1280736 |
500 g |
Each for $184.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-99-1 | |
201.562 | |
DFXQXFGFOLXAPO-UHFFFAOYSA-N | |
7320 |
C7H4ClNO4 | |
MFCD00007079 | |
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene | |
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl |
Specifications
96-99-1 | |
1.645 | |
C7H4ClNO4 | |
500 g | |
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene | |
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl | |
7320 | |
99% |
180°C to 184°C | |
Odorless | |
MFCD00007079 | |
783626 | |
DFXQXFGFOLXAPO-UHFFFAOYSA-N | |
201.562 | |
201.57 | |
4-Chloro-3-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-550-9
RTECSNumber : DG5425050
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only