missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrophenol, 99%, Thermo Scientific Chemicals

$75.04 - $246.53

Chemical Identifiers

CAS 610-78-6
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043546
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
PubChem CID 69127
IUPAC Name 4-chloro-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1010806
View Documents
Thermo Scientific Chemicals
L1010806
5 g
Each for $75.04
 
AAL1010814
View Documents
Thermo Scientific Chemicals
L1010814
25 g
Each for $246.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

610-78-6
173.552
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
C6H4ClNO3
MFCD00043546
69127
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl

Specifications

610-78-6
Characteristic
MFCD00043546
2094546
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
69127
99%
124°C to 129°C
C6H4ClNO3
5 g
phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
173.552
173.56
4-Chloro-3-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only