missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrophenol 98.0+%, TCI America™

$54.06 - $188.27

Chemical Identifiers

CAS 610-78-6
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043546
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
Synonym phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
PubChem CID 69127
IUPAC Name 4-chloro-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C15295G
View Documents
TCI America
C15295G
5 g
Each for $54.06
Only null left
 
C152925G
View Documents
TCI America
C152925G
25 g
Each for $188.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

610-78-6
173.552
JUIKCULGDIZNDI-UHFFFAOYSA-N
69127
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
C6H4ClNO3
MFCD00043546
phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
4-chloro-3-nitrophenol

Specifications

610-78-6
C6H4ClNO3
5 g
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
69127
≥98.0% (GC,T)
4-Chloro-3-nitrophenol
127°C
MFCD00043546
phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
173.552
173.55
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'