missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrotoluene, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1138336

 View more versions of this product

Catalog No. AAA1138336


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-3-nitrotoluene
∼7°C
260°C to 262°C
C7H6ClNO2
MFCD00007085
UN2433
4-chloro-3-nitrotoluene, 3-nitro-4-chlorotoluene, benzene, 1-chloro-4-methyl-2-nitro, 2-chloro-5-methylnitrobenzene, 1-chloro-4-methyl-2-nitro-benzene, toluene, 4-chloro-3-nitro, 3-nitro-4-chlorotoluol, pubchem19752, 4,3-chloronitrotoluene, 4-chloro-3-nitro-toluene
CC1=CC=C(Cl)C(=C1)[N+]([O-])=O
171.58
171.58
89-60-1
1.295
129°C (264°F)
1.558
500 g
511055
NWESJZZPAJGHRZ-UHFFFAOYSA-N
1-chloro-4-methyl-2-nitrobenzene
6976
≥97%

Chemical Identifiers

89-60-1
171.58
NWESJZZPAJGHRZ-UHFFFAOYSA-N
6976
CC1=CC=C(Cl)C(=C1)[N+]([O-])=O
C7H6ClNO2
MFCD00007085
4-chloro-3-nitrotoluene, 3-nitro-4-chlorotoluene, benzene, 1-chloro-4-methyl-2-nitro, 2-chloro-5-methylnitrobenzene, 1-chloro-4-methyl-2-nitro-benzene, toluene, 4-chloro-3-nitro, 3-nitro-4-chlorotoluol, pubchem19752, 4,3-chloronitrotoluene, 4-chloro-3-nitro-toluene
1-chloro-4-methyl-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, LIQUID

EINECSNumber : 201-922-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only