missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-4'-hydroxybenzophenone 98.0+%, TCI America™
$124.89 - $965.59
Chemical Identifiers
| CAS | 42019-78-3 |
|---|---|
| Molecular Formula | C13H9ClO2 |
| Molecular Weight (g/mol) | 232.66 |
| MDL Number | MFCD00002357 |
| InChI Key | RUETVLNXAGWCDS-UHFFFAOYSA-N |
| Synonym | 4-chloro-4'-hydroxybenzophenone, 4-chlorophenyl 4-hydroxyphenyl methanone, 4-hydroxy-4'-chlorobenzophenone, 4-p-chlorobenzoylphenol, 4-4-chlorobenzoyl phenol, 4-chlorophenyl 4-hydroxyphenyl-methanone, methanone, 4-chlorophenyl 4-hydroxyphenyl, 4-chlorophenyl-4-hydroxyphenyl methanone, 4-chlorophenyl 4-hydroxyphenyl ketone, pubchem3317 |
| PubChem CID | 123504 |
| IUPAC Name | 4-(4-chlorobenzoyl)phenol |
| SMILES | OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 42019-78-3 | |
| 232.66 | |
| RUETVLNXAGWCDS-UHFFFAOYSA-N | |
| 123504 | |
| OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1 |
| C13H9ClO2 | |
| MFCD00002357 | |
| 4-chloro-4'-hydroxybenzophenone, 4-chlorophenyl 4-hydroxyphenyl methanone, 4-hydroxy-4'-chlorobenzophenone, 4-p-chlorobenzoylphenol, 4-4-chlorobenzoyl phenol, 4-chlorophenyl 4-hydroxyphenyl-methanone, methanone, 4-chlorophenyl 4-hydroxyphenyl, 4-chlorophenyl-4-hydroxyphenyl methanone, 4-chlorophenyl 4-hydroxyphenyl ketone, pubchem3317 | |
| 4-(4-chlorobenzoyl)phenol |
Specifications
| 42019-78-3 | |
| C13H9ClO2 | |
| 25 g | |
| RUETVLNXAGWCDS-UHFFFAOYSA-N | |
| 4-(4-chlorobenzoyl)phenol | |
| 123504 | |
| ≥98.0% (GC) | |
| 4-Chloro-4′-hydroxybenzophenone |
| 180°C | |
| MFCD00002357 | |
| 4-chloro-4'-hydroxybenzophenone, 4-chlorophenyl 4-hydroxyphenyl methanone, 4-hydroxy-4'-chlorobenzophenone, 4-p-chlorobenzoylphenol, 4-4-chlorobenzoyl phenol, 4-chlorophenyl 4-hydroxyphenyl-methanone, methanone, 4-chlorophenyl 4-hydroxyphenyl, 4-chlorophenyl-4-hydroxyphenyl methanone, 4-chlorophenyl 4-hydroxyphenyl ketone, pubchem3317 | |
| OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1 | |
| 232.66 | |
| 232.66 | |
| Crystalline Powder |
Safety and Handling
TSCA : No