missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00007EA

 View more versions of this product

Catalog No. AW00007EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine
94.5
95%
MFCD00174019
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
252.73
252.75
189681-04-7
100.0
C10H5ClN2S2
10g
VNWBUFWVNCUUKY-UHFFFAOYSA-N
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
735732
95%

Chemical Identifiers

189681-04-7
252.73
VNWBUFWVNCUUKY-UHFFFAOYSA-N
735732
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
C10H5ClN2S2
MFCD00174019
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
Videos
SDS Documents

Documents

Promotions

Promotions