missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™
$102.52 - $350.44
Chemical Identifiers
| CAS | 116941-51-6 |
|---|---|
| Molecular Formula | C24H17Cl |
| Molecular Weight (g/mol) | 340.85 |
| InChI Key | FZOIFXXPBLTVIG-UHFFFAOYSA-N |
| Synonym | 5′C-(4-Chlorophenyl)-1,1′C:3′C,1′C′C-terphenyl |
| PubChem CID | 11110599 |
| IUPAC Name | 1-(4-chlorophenyl)-3,5-diphenylbenzene |
| SMILES | C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C3008200MG
|
TCI America
C3008200MG |
200 mg |
Each for $102.52
|
|
|||||
|
C30081G
|
TCI America
C30081G |
1 g |
Each for $350.44
|
|
|||||
Chemical Identifiers
| 116941-51-6 | |
| 340.85 | |
| 5′C-(4-Chlorophenyl)-1,1′C:3′C,1′C′C-terphenyl | |
| 1-(4-chlorophenyl)-3,5-diphenylbenzene |
| C24H17Cl | |
| FZOIFXXPBLTVIG-UHFFFAOYSA-N | |
| 11110599 | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4 |
Specifications
| 116941-51-6 | |
| White | |
| 200 mg | |
| FZOIFXXPBLTVIG-UHFFFAOYSA-N | |
| 1-(4-chlorophenyl)-3,5-diphenylbenzene | |
| 11110599 | |
| ≥98.0% (GC) | |
| 4-Chloro-5′-phenyl-1,1′:3′,1′′-terphenyl |
| 143°C | |
| C24H17Cl | |
| 5′C-(4-Chlorophenyl)-1,1′C:3′C,1′C′C-terphenyl | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4 | |
| 340.85 | |
| 340.85 | |
| Crystalline Powder |
Safety and Handling
TSCA : No