missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-N-(3-methylphenyl)butyramide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 73863-44-2
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD00086969
InChI Key GBYXVHYDSWAERN-UHFFFAOYSA-N
Synonym 4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
PubChem CID 1809759
IUPAC Name 4-chloro-N-(3-methylphenyl)butanamide
SMILES CC1=CC=CC(NC(=O)CCCCl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH58249MC
View Documents
Thermo Scientific Chemicals
H58249MC
100mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH58249ME
View Documents
Thermo Scientific Chemicals
H58249ME
500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

73863-44-2
211.69
GBYXVHYDSWAERN-UHFFFAOYSA-N
1809759
CC1=CC=CC(NC(=O)CCCCl)=C1
C11H14ClNO
MFCD00086969
4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
4-chloro-N-(3-methylphenyl)butanamide

Specifications

73863-44-2
MFCD00086969
4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
CC1=CC=CC(NC(=O)CCCCl)=C1
211.69
211.69
4-Chloro-N-(3-methylphenyl)butyramide
C11H14ClNO
100mg
GBYXVHYDSWAERN-UHFFFAOYSA-N
4-chloro-N-(3-methylphenyl)butanamide
1809759
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'