missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-N-methylaniline 97.0+%, TCI America™
Supplier: TCI America C162025G
Specifications
4-Chloro-N-methylaniline | |
142°C | |
MFCD00000614 | |
1993 | |
XCEYKKJMLOFDSS-UHFFFAOYSA-N | |
4-chloro-N-methylaniline | |
70272 | |
≥97.0% (GC,T) |
932-96-7 | |
C7H8ClN | |
25 g | |
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine | |
CNC1=CC=C(C=C1)Cl | |
141.598 | |
141.60 | |
Liquid |
Chemical Identifiers
932-96-7 | |
141.598 | |
XCEYKKJMLOFDSS-UHFFFAOYSA-N | |
70272 | |
CNC1=CC=C(C=C1)Cl |
C7H8ClN | |
MFCD00000614 | |
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine | |
4-chloro-N-methylaniline |
Safety and Handling
RTECSNumber : CX9857900
TSCA : No