Learn More
4-Chloroaniline hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1374406
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chloroaniline hydrochloride | |
>230°C (sublimation) | |
C6H7Cl2N | |
5 g | |
3560964 | |
ISJBQSJDQZLCSF-UHFFFAOYSA-N | |
4-chloroaniline;hydrochloride | |
5284363 | |
97% |
20265-96-7 | |
Characteristic | |
MFCD00042057 | |
UN2018 | |
4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride | |
C1=CC(=CC=C1N)Cl.Cl | |
164.029 | |
164.03 |
Chemical Identifiers
20265-96-7 | |
164.029 | |
ISJBQSJDQZLCSF-UHFFFAOYSA-N | |
5284363 | |
C1=CC(=CC=C1N)Cl.Cl |
C6H7Cl2N | |
MFCD00042057 | |
4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride | |
4-chloroaniline;hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
EINECSNumber : 243-656-5
RTECSNumber : BX1394000
TSCA : Yes
Recommended Storage : Ambient temperatures