missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamide, 98+%

CAS: 619-56-7 | C7H6ClNO | 155.58 g/mol

$102.03 - $1,311.62

Chemical Identifiers

CAS 619-56-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007993
InChI Key BLNVISNJTIRAHF-UHFFFAOYSA-N
Synonym p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
PubChem CID 12084
IUPAC Name 4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1380909
View Documents
Thermo Scientific Chemicals
A1380909
10 g N/A
Only null left
 
AAA1380918
View Documents
Thermo Scientific Chemicals
A1380918
50 g N/A
Only null left
 
AAA1380930
View Documents
Thermo Scientific Chemicals
A1380930
250 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-56-7
155.58
BLNVISNJTIRAHF-UHFFFAOYSA-N
12084
NC(=O)C1=CC=C(Cl)C=C1
C7H6ClNO
MFCD00007993
p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
4-chlorobenzamide

Specifications

619-56-7
C7H6ClNO
10 g
p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
NC(=O)C1=CC=C(Cl)C=C1
155.58
155.58
4-Chlorobenzamide
176°C to 180°C
MFCD00007993
774622
BLNVISNJTIRAHF-UHFFFAOYSA-N
4-chlorobenzamide
12084
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 210-601-1

RTECSNumber : CV2443811

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only