Learn More
4-Chlorobenzenesulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 158565000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chlorobenzenesulfonyl chloride | |
98-60-2 | |
100.0 | |
Authentic | |
Plastic bottle | |
ClC6H4SO2Cl | |
500g | |
4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride | |
ZLYBFBAHAQEEQQ-UHFFFAOYSA-N | |
4-chlorobenzenesulfonyl chloride | |
7398 | |
97% |
97% | |
96.0 | |
107°C | |
97% | |
C6H4Cl2O2S | |
MFCD00007439 | |
11, 55 | |
Solubility in water: insoluble in water | |
ClC1=CC=C(C=C1)S(Cl)(=O)=O | |
211.06 | |
211.07 |
Chemical Identifiers
98-60-2 | |
211.06 | |
ZLYBFBAHAQEEQQ-UHFFFAOYSA-N | |
7398 | |
ClC1=CC=C(C=C1)S(Cl)(=O)=O |
C6H4Cl2O2S | |
MFCD00007439 | |
4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride | |
4-chlorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-685-3
RTECSNumber : DB8925000
TSCA : TSCA
RUO â Research Use Only