missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108730250

 View more versions of this product

Catalog No. AC108730250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chlorobenzotrifluoride
98-56-6
100.0
Colorless
136.0°C to 138.0°C
Authentic
Glass bottle
1.4453 to 1.4473
MFCD00000627
1.35
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
C1=CC(=CC=C1C(F)(F)F)Cl
180.56
0.67 mPa.s (38°C)
98%
98%
97.5
-36.0°C
1.3500g/mL
47°C
98%
C7H4ClF3
ClC6H4CF3
25 mL
15, 2127
QULYNCCPRWKEMF-UHFFFAOYSA-N
1-chloro-4-(trifluoromethyl)benzene
7394
180.56
Liquid

Chemical Identifiers

98-56-6
180.56
QULYNCCPRWKEMF-UHFFFAOYSA-N
7394
C1=CC(=CC=C1C(F)(F)F)Cl
C7H4ClF3
MFCD00000627
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
1-chloro-4-(trifluoromethyl)benzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou

GHS Signal Word: Danger

EINECSNumber : 202-681-1

RTECSNumber : XS9145000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only