Learn More
4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 108732500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chlorobenzotrifluoride | |
98-56-6 | |
100.0 | |
Colorless | |
136.0°C to 138.0°C | |
Authentic | |
Glass bottle | |
1.4453 to 1.4473 | |
MFCD00000627 | |
1.35 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
C1=CC(=CC=C1C(F)(F)F)Cl | |
180.56 | |
0.67 mPa.s (38°C) | |
98% |
98% | |
97.5 | |
-36.0°C | |
1.3500g/mL | |
47°C | |
98% | |
C7H4ClF3 | |
ClC6H4CF3 | |
250 mL | |
15, 2127 | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
1-chloro-4-(trifluoromethyl)benzene | |
7394 | |
180.56 | |
Liquid |
Chemical Identifiers
98-56-6 | |
180.56 | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
7394 | |
C1=CC(=CC=C1C(F)(F)F)Cl |
C7H4ClF3 | |
MFCD00000627 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
1-chloro-4-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou
GHS Signal Word: Danger
EINECSNumber : 202-681-1
RTECSNumber : XS9145000
TSCA : TSCA
RUO – Research Use Only