missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoyl chloride, 98%

CAS: 122-01-0 | C7H4Cl2O | 175.008 g/mol

$66.03 - $192.42

Chemical Identifiers

CAS 122-01-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00000686
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
PubChem CID 8501
ChEBI CHEBI:60716
IUPAC Name 4-chlorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1632522
View Documents
Thermo Scientific Chemicals
A1632522
100 g
Each for $66.03
Only null left
 
AAA1632536
View Documents
Thermo Scientific Chemicals
A1632536
500 g
Each for $192.42
Only null left
 
Description

Description

4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

Solubility
Reacts with water. Reacts with alcohol.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alcohol, bases.
Specifications

Chemical Identifiers

122-01-0
175.008
RKIDDEGICSMIJA-UHFFFAOYSA-N
8501
4-chlorobenzoyl chloride
C7H4Cl2O
MFCD00000686
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
CHEBI:60716
C1=CC(=CC=C1C(=O)Cl)Cl

Specifications

122-01-0
1.377
105°C (221°F)
C7H4Cl2O
MFCD00000686
UN3265
Moisture sensitive
Reacts with water. Reacts with alcohol.
C1=CC(=CC=C1C(=O)Cl)Cl
175.008
CHEBI:60716
98%
12°C to 14°C
221°C to 222°C
Acrid
1.578
100 g
471606
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
RKIDDEGICSMIJA-UHFFFAOYSA-N
4-chlorobenzoyl chloride
8501
175.01
4-Chlorobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 204-515-3

RTECSNumber : DM6635510

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only