Learn More
4-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals
$44.19 - $1296.59
Chemical Identifiers
CAS | 104-83-6 |
---|---|
Molecular Formula | C7H6Cl2 |
Molecular Weight (g/mol) | 161.025 |
MDL Number | MFCD00000914 |
InChI Key | JQZAEUFPPSRDOP-UHFFFAOYSA-N |
Synonym | 4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro |
PubChem CID | 7723 |
IUPAC Name | 1-chloro-4-(chloromethyl)benzene |
SMILES | C1=CC(=CC=C1CCl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1469614
|
Thermo Scientific Chemicals
A1469614 |
25 g |
Each for $44.19
|
|
||||||
AAA1469630
|
Thermo Scientific Chemicals
A1469630 |
250 g |
Each for $100.07
|
|
||||||
AAA146960B
|
Thermo Scientific Chemicals
A146960B |
1000 g |
Each for $328.71
|
|
||||||
AAA146960I
|
Thermo Scientific Chemicals
A146960I |
5000 g |
Each for $1,296.59
|
|
||||||
Description
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
104-83-6 | |
161.025 | |
JQZAEUFPPSRDOP-UHFFFAOYSA-N | |
7723 | |
C1=CC(=CC=C1CCl)Cl |
C7H6Cl2 | |
MFCD00000914 | |
4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro | |
1-chloro-4-(chloromethyl)benzene |
Specifications
104-83-6 | |
1.275 | |
107°C (225°F) | |
C7H6Cl2 | |
MFCD00000914 | |
UN3427 | |
4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro | |
JQZAEUFPPSRDOP-UHFFFAOYSA-N | |
1-chloro-4-(chloromethyl)benzene | |
7723 | |
≥98% |
26°C to 30°C | |
220°C to 221°C | |
Pungent | |
1.5575 | |
25 g | |
471558 | |
Insoluble in water. | |
C1=CC(=CC=C1CCl)Cl | |
161.025 | |
161.03 | |
4-Chlorobenzyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
H302+H312+H332-H317
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROBENZYL CHLORIDES, SOLID
EINECSNumber : 203-242-7
RTECSNumber : XT0720000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only