Learn More
4-Chlorocinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals
$120.52 - $120.52
Chemical Identifiers
CAS | 1615-02-7 |
---|---|
Molecular Formula | C9H7ClO2 |
Molecular Weight (g/mol) | 182.60 |
MDL Number | MFCD00004396 |
InChI Key | GXLIFJYFGMHYDY-ZZXKWVIFSA-N |
Synonym | 4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid |
PubChem CID | 637797 |
ChEBI | CHEBI:61116 |
IUPAC Name | (E)-3-(4-chlorophenyl)prop-2-enoic acid |
SMILES | OC(=O)\C=C\C1=CC=C(Cl)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC108940250
|
Thermo Scientific Chemicals
108940250 |
25 g | Plastic bottle |
Each for $120.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1615-02-7 | |
182.60 | |
GXLIFJYFGMHYDY-ZZXKWVIFSA-N | |
637797 | |
(E)-3-(4-chlorophenyl)prop-2-enoic acid |
C9H7ClO2 | |
MFCD00004396 | |
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid | |
CHEBI:61116 | |
OC(=O)\C=C\C1=CC=C(Cl)C=C1 |
Specifications
1615-02-7 | |
100.0 | |
White | |
99% | |
C9H7ClO2 | |
MFCD00004396 | |
09, 594 | |
GXLIFJYFGMHYDY-ZZXKWVIFSA-N | |
(E)-3-(4-chlorophenyl)prop-2-enoic acid | |
637797 | |
182.61 | |
Fine Crystalline Powder |
98.5 | |
246°C to 250°C | |
Authentic | |
Plastic bottle | |
ClC6H4CH=CHCO2H | |
25 g | |
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid | |
OC(=O)\C=C\C1=CC=C(Cl)C=C1 | |
182.60 | |
CHEBI:61116 | |
99% | |
4-Chlorocinnamic acid, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 216-564-8
RTECSNumber : GD8490000
RUO – Research Use Only