missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Chloromethyl)benzoic acid, 96%, Thermo Scientific Chemicals

$118.18 - $351.89

Chemical Identifiers

CAS 1642-81-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00002568
InChI Key OITNBJHJJGMFBN-UHFFFAOYSA-N
Synonym 4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
PubChem CID 74234
IUPAC Name 4-(chloromethyl)benzoic acid
SMILES C1=CC(=CC=C1CCl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2227506
View Documents
Thermo Scientific Chemicals
B2227506
5 g
Each for $118.18
 
AAB2227514
View Documents
Thermo Scientific Chemicals
B2227514
25 g
Each for $351.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1642-81-5
170.592
OITNBJHJJGMFBN-UHFFFAOYSA-N
74234
C1=CC(=CC=C1CCl)C(=O)O
C8H7ClO2
MFCD00002568
4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
4-(chloromethyl)benzoic acid

Specifications

1642-81-5
C8H7ClO2
5 g
1907970
OITNBJHJJGMFBN-UHFFFAOYSA-N
4-(chloromethyl)benzoic acid
74234
96%
199°C to 202°C
MFCD00002568
UN3261
4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
C1=CC(=CC=C1CCl)C(=O)O
170.592
170.6
4-(Chloromethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 216-697-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only