missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Chloromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

$127.55 - $1649.45

Chemical Identifiers

CAS 874-86-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00019761
InChI Key LOQLDQJTSMKBJU-UHFFFAOYSA-N
Synonym 4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
PubChem CID 70127
IUPAC Name 4-(chloromethyl)benzonitrile
SMILES C1=CC(=CC=C1CCl)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2213809
View Documents
Thermo Scientific Chemicals
B2213809
10 g
Each for $127.55
 
AAB2213818
View Documents
Thermo Scientific Chemicals
B2213818
50 g
Each for $416.03
 
AAB2213830
View Documents
Thermo Scientific Chemicals
B2213830
250 g
Each for $1,649.45
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

874-86-2
151.593
LOQLDQJTSMKBJU-UHFFFAOYSA-N
70127
C1=CC(=CC=C1CCl)C#N
C8H6ClN
MFCD00019761
4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
4-(chloromethyl)benzonitrile

Specifications

874-86-2
C8H6ClN
10 g
4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
C1=CC(=CC=C1CCl)C#N
151.593
151.6
4-(Chloromethyl)benzonitrile
77°C to 80°C
MFCD00019761
UN2923
LOQLDQJTSMKBJU-UHFFFAOYSA-N
4-(chloromethyl)benzonitrile
70127
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c

H301+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 212-869-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only