missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Chloromethyl)benzyl alcohol, 97%, Thermo Scientific Chemicals
$81.30 - $306.86
Chemical Identifiers
CAS | 16473-35-1 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.61 |
InChI Key | OGALXJIOJZXBBP-UHFFFAOYSA-N |
Synonym | 4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride |
PubChem CID | 5107660 |
IUPAC Name | [4-(chloromethyl)phenyl]methanol |
SMILES | C1=CC(=CC=C1CO)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC438920010
|
Thermo Scientific Chemicals
438920010 |
1 g |
Each for $81.30
|
|
|||||
AC438920050
|
Thermo Scientific Chemicals
438920050 |
5 g |
Each for $306.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16473-35-1 | |
156.61 | |
4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride | |
[4-(chloromethyl)phenyl]methanol |
C8H9ClO | |
OGALXJIOJZXBBP-UHFFFAOYSA-N | |
5107660 | |
C1=CC(=CC=C1CO)CCl |
Specifications
16473-35-1 | |
Authentic | |
Glass bottle | |
ClCH2C6H4CH2OH | |
4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride | |
C1=CC(=CC=C1CO)CCl | |
156.61 | |
156.61 | |
4-(Chloromethyl)benzyl alcohol |
58.0°C to 60.0°C | |
96% min. (GC) | |
C8H9ClO | |
1 g | |
OGALXJIOJZXBBP-UHFFFAOYSA-N | |
[4-(chloromethyl)phenyl]methanol | |
5107660 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only