missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenacyl Bromide 98.0+%, TCI America™

Supplier:  TCI America B0568500G

 View more versions of this product

Catalog No. B0568500G


Explore available promotions

Description

Description

Specifications

Specifications

4-Chlorophenacyl Bromide
99°C
C8H6BrClO
500 g
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
C1=CC(=CC=C1C(=O)CBr)Cl
233.489
233.49
Crystalline Powder
536-38-9
White-Yellow
MFCD00000625
3261
FLAYZKKEOIAALB-UHFFFAOYSA-N
2-bromo-1-(4-chlorophenyl)ethanone
68303
≥98.0% (GC,T)

Chemical Identifiers

536-38-9
233.489
FLAYZKKEOIAALB-UHFFFAOYSA-N
68303
C1=CC(=CC=C1C(=O)CBr)Cl
C8H6BrClO
MFCD00000625
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
2-bromo-1-(4-chlorophenyl)ethanone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2934

RTECSNumber : AM5978800

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions