missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A156020B

 View more versions of this product

Catalog No. AAA156020B


Explore available promotions

Description

Description

It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

Solubility
Soluble in water (27g/L).

Notes
Air sensitive. Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-Chlorophenol
39°C to 43°C
217°C to 220°C
Phenol-like
MFCD00002318
UN2020
Air Sensitive
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
4-chlorophenol
4684
128.56
106-48-9
1.306
115°C (239°F)
C6H5ClO
1000 g
507004
14,2154
Soluble in water (27g/L).
C1=CC(=CC=C1O)Cl
128.555
CHEBI:28078
99%

Chemical Identifiers

106-48-9
128.555
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
4684
4-chlorophenol
C6H5ClO
MFCD00002318
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
CHEBI:28078
C1=CC(=CC=C1O)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

EINECSNumber : 203-402-6

RTECSNumber : SK2800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only