missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenoxyacetonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1538914

 View more versions of this product

Catalog No. AAA1538914


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chlorophenoxyacetonitrile
3598-13-8
25 g
MFCD00031535
2442163
YUGDKEWUYZXXRU-UHFFFAOYSA-N
2-(4-chlorophenoxy)acetonitrile
306444
98%
45°C to 49°C
149°C to 150°C (15 mmHg)
C8H6ClNO
UN3439
2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
ClC1=CC=C(OCC#N)C=C1
167.59
167.6

Chemical Identifiers

3598-13-8
167.59
YUGDKEWUYZXXRU-UHFFFAOYSA-N
306444
ClC1=CC=C(OCC#N)C=C1
C8H6ClNO
MFCD00031535
2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
2-(4-chlorophenoxy)acetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only