Learn More
(4-Chlorophenyl)(4-piperidyl)methanone hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CD10000CB
Specifications
(4-Chlorophenyl)(4-piperidyl)methanone hydrochloride | |
94.5 | |
95% | |
C12H15ClNO | |
250mg | |
IYGWDOXHCPQXKN-UHFFFAOYSA-O | |
(4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride | |
2777114 | |
95% |
55695-51-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD00053028 | |
4-chlorophenyl piperidin-4-yl methanone hydrochloride, 4-4-chlorobenzoyl piperidine hydrochloride, 4-chlorophenyl 4-piperidyl methanone hydrochloride, 4-chlorophenyl 4-piperidinyl methanone hydrochloride, 4-p-chlorobenzoyl piperidine hydrochloride, 4-4-chlorophenyl carbonyl piperidine hydrochloride, methanone, 4-chlorophenyl-4-piperidinyl-, hydrochloride, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, bestipharma 579-258, 4-4-chlorobenzoyl piperidine hcl | |
ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1 | |
224.71 | |
260.16 |
Chemical Identifiers
55695-51-7 | |
224.71 | |
IYGWDOXHCPQXKN-UHFFFAOYSA-O | |
2777114 | |
ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1 |
C12H15ClNO | |
MFCD00053028 | |
4-chlorophenyl piperidin-4-yl methanone hydrochloride, 4-4-chlorobenzoyl piperidine hydrochloride, 4-chlorophenyl 4-piperidyl methanone hydrochloride, 4-chlorophenyl 4-piperidinyl methanone hydrochloride, 4-p-chlorobenzoyl piperidine hydrochloride, 4-4-chlorophenyl carbonyl piperidine hydrochloride, methanone, 4-chlorophenyl-4-piperidinyl-, hydrochloride, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, bestipharma 579-258, 4-4-chlorobenzoyl piperidine hcl | |
(4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning