missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 195062-61-4
Molecular Formula C12H16BClO2
Molecular Weight (g/mol) 238.52
MDL Number MFCD05663875
InChI Key NYARTXMDWRAVIX-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
PubChem CID 10633712
IUPAC Name 2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444690050
SDS View Documents
Thermo Scientific Chemicals
444690050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444690250
SDS View Documents
Thermo Scientific Chemicals
444690250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

195062-61-4
238.52
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
C12H16BClO2
MFCD05663875
2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

195062-61-4
96% min. (GC)
C12H16BClO2
5g
NYARTXMDWRAVIX-UHFFFAOYSA-N
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10633712
97%
>110°C
Glass bottle
MFCD05663875
2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
238.52
238.52
4-Chlorophenylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only