missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals

$162.08 - $162.08

Chemical Identifiers

CAS 10130-89-9
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.63
InChI Key PTCSSXYPZOFISK-UHFFFAOYSA-N
Synonym 4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
PubChem CID 82385
IUPAC Name 4-chlorosulfonylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC405060050
SDS View Documents
Thermo Scientific Chemicals
405060050
5 g Glass bottle
Each for $162.08
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10130-89-9
220.63
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
4-chlorosulfonylbenzoic acid
C7H5ClO4S
PTCSSXYPZOFISK-UHFFFAOYSA-N
82385
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl

Specifications

10130-89-9
White to Beige
96%
C7H5ClO4S
5 g
Solubility in water: decomposes.
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl
220.63
220.63
Powder
230.0°C
Authentic
Glass bottle
CO2HC6H4SO2Cl
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
PTCSSXYPZOFISK-UHFFFAOYSA-N
4-chlorosulfonylbenzoic acid
82385
96%
4-(Chlorosulfonyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 233-367-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only