missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chlorotetrahydropyran, 96%, Thermo Scientific™

Chemical Identifiers

CAS 1768-64-5
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
InChI Key DHRSKOBIDIDMJZ-UHFFFAOYSA-N
Synonym 4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene
PubChem CID 137202
IUPAC Name 4-chlorooxane
SMILES C1COCCC1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434270010
View Documents
Thermo Scientific Chemicals
434270010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434270050
View Documents
Thermo Scientific Chemicals
434270050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1768-64-5
120.58
4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene
4-chlorooxane
C5H9ClO
DHRSKOBIDIDMJZ-UHFFFAOYSA-N
137202
C1COCCC1Cl

Specifications

1768-64-5
100.0
150°C
Authentic
Glass bottle
1g
4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene
C1COCCC1Cl
120.58
120.58
4-chlorotetrahydropyran
95.0
1.1200g/mL
46°C
95% min. (GC)
C5H9ClO
1.12
DHRSKOBIDIDMJZ-UHFFFAOYSA-N
4-chlorooxane
137202
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapour.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only