missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyano-3-fluoropyridine, 95%, Thermo Scientific Chemicals

$294.96 - $1011.48

Chemical Identifiers

CAS 113770-88-0
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD04112524
InChI Key RFQHDTRSEWCXJP-UHFFFAOYSA-N
PubChem CID 22156440
IUPAC Name 3-fluoropyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6159403
View Documents
Thermo Scientific Chemicals
H6159403
1 g
Each for $294.96
 
AAH6159406
View Documents
Thermo Scientific Chemicals
H6159406
5 g
Each for $1,011.48
 
Description

Description

4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

113770-88-0
122.102
RFQHDTRSEWCXJP-UHFFFAOYSA-N
3-fluoropyridine-4-carbonitrile
C6H3FN2
MFCD04112524
22156440
C1=CN=CC(=C1C#N)F

Specifications

113770-88-0
C6H3FN2
1 g
3-fluoro-4-cyanopyridine, 3-fluoroisonicotinonitrile, 3-fluoro-4-cyano-pyridine, 3-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,3-fluoro, 4-pyridinecarbonitrile, 3-fluoro, 4-pyridinecarbonitrile,3-fluoro-9ci, 3-fluoro-4-pyridinecarbonitrile, 3-fluoranylpyridine-4-carbonitrile, 4-pyridinecarbonitrile, 3-fluoro-9ci
RFQHDTRSEWCXJP-UHFFFAOYSA-N
3-fluoropyridine-4-carbonitrile
22156440
95%
4-Cyano-3-fluoropyridine
White to Yellow
MFCD04112524
UN3439
Slightly soluble in water.
C1=CN=CC(=C1C#N)F
122.102
122.1
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only