missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Cyano-4-decyloxybiphenyl 98.0+%, TCI America™
$331.79 - $983.79
Chemical Identifiers
CAS | 70247-25-5 |
---|---|
Molecular Formula | C23H29NO |
Molecular Weight (g/mol) | 335.491 |
MDL Number | MFCD01218034 |
InChI Key | VCYXELFOIWRYLA-UHFFFAOYSA-N |
PubChem CID | 112405 |
IUPAC Name | 4-(4-decoxyphenyl)benzonitrile |
SMILES | CCCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N |
Chemical Identifiers
70247-25-5 | |
335.491 | |
VCYXELFOIWRYLA-UHFFFAOYSA-N | |
4-(4-decoxyphenyl)benzonitrile |
C23H29NO | |
MFCD01218034 | |
112405 | |
CCCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N |
Specifications
70247-25-5 | |
C23H29NO | |
1 g | |
VCYXELFOIWRYLA-UHFFFAOYSA-N | |
4-(4-decoxyphenyl)benzonitrile | |
112405 | |
≥98.0% (GC) | |
4′-Cyano-4-decyloxybiphenyl |
White | |
MFCD01218034 | |
4′C-Decyloxy-4-biphenylcarbonitrile, 4′C-(Decyloxy)-[1,1′C-biphenyl]-4-carbonitrile, 10OCB | |
CCCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N | |
335.491 | |
335.49 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes