Learn More
4-Cyano-4″-n-pentyl-p-terphenyl, 99%, Thermo Scientific Chemicals
$127.10 - $458.37
Chemical Identifiers
CAS | 54211-46-0 |
---|---|
Molecular Formula | C24H23N |
Molecular Weight (g/mol) | 325.455 |
MDL Number | MFCD00799423 |
InChI Key | AITQOXOBSMXBRV-UHFFFAOYSA-N |
Synonym | 4-pentyl-1,1':4',1-terphenyl-4-carbonitrile, 4-cyano-4-n-pentyl-p-terphenyl, 4-cyano-4-pentyl-p-terphenyl, 1,1':4',1-terphenyl-4-carbonitrile, 4-pentyl, 4-4-4-pentylphenyl phenyl benzonitrile, pubchem9116, 4-pentyl-p-terphenyl-4-carbonitrile, 4-cyano-4'-pentyl-p-terphenyl |
PubChem CID | 104707 |
IUPAC Name | 4-[4-(4-pentylphenyl)phenyl]benzonitrile |
SMILES | CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2260702
|
Thermo Scientific Chemicals
B2260702 |
0.5 g |
Each for $127.10
|
|
|||||
AAB2260788
|
Thermo Scientific Chemicals
B2260788 |
2.5 g |
Each for $458.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54211-46-0 | |
325.455 | |
AITQOXOBSMXBRV-UHFFFAOYSA-N | |
104707 | |
CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N |
C24H23N | |
MFCD00799423 | |
4-pentyl-1,1':4',1-terphenyl-4-carbonitrile, 4-cyano-4-n-pentyl-p-terphenyl, 4-cyano-4-pentyl-p-terphenyl, 1,1':4',1-terphenyl-4-carbonitrile, 4-pentyl, 4-4-4-pentylphenyl phenyl benzonitrile, pubchem9116, 4-pentyl-p-terphenyl-4-carbonitrile, 4-cyano-4'-pentyl-p-terphenyl | |
4-[4-(4-pentylphenyl)phenyl]benzonitrile |
Specifications
54211-46-0 | |
C24H23N | |
0.5 g | |
4-pentyl-1,1':4',1-terphenyl-4-carbonitrile, 4-cyano-4-n-pentyl-p-terphenyl, 4-cyano-4-pentyl-p-terphenyl, 1,1':4',1-terphenyl-4-carbonitrile, 4-pentyl, 4-4-4-pentylphenyl phenyl benzonitrile, pubchem9116, 4-pentyl-p-terphenyl-4-carbonitrile, 4-cyano-4'-pentyl-p-terphenyl | |
CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N | |
325.455 | |
325.45 | |
4-Cyano-4''-n-pentyl-p-terphenyl |
128°C to 130°C | |
MFCD00799423 | |
UN3439 | |
AITQOXOBSMXBRV-UHFFFAOYSA-N | |
4-[4-(4-pentylphenyl)phenyl]benzonitrile | |
104707 | |
99% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 259-028-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only