missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyanobenzophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1576403

 View more versions of this product

Catalog No. AAA1576403


Explore available promotions

Description

Description

Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

4-Cyanobenzophenone
110°C to 114°C
MFCD00016382
UN3439
4-cyanobenzophenone, 4-benzoyl-benzonitrile, benzonitrile, 4-benzoyl, 4-cyano-benzophenon, 4-cyano-benzophenone, acmc-20amk8, #, benzonitrile,4-benzoyl, 4-phenylcarbonyl benzenecarbonitrile
YSZWJJANSNFQMM-UHFFFAOYSA-N
4-benzoylbenzonitrile
73921
98%
1503-49-7
C14H9NO
1 g
641181
Insoluble in water.
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N
207.232
207.23

Chemical Identifiers

1503-49-7
207.232
YSZWJJANSNFQMM-UHFFFAOYSA-N
4-benzoylbenzonitrile
C14H9NO
MFCD00016382
73921
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 216-126-6

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only