missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyanophenylacetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5462-71-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD06798066
InChI Key WEBXRQONNWEETE-UHFFFAOYSA-N
Synonym 2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
PubChem CID 79587
IUPAC Name 2-(4-cyanophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437730010
SDS View Documents
Thermo Scientific Chemicals
437730010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437730050
SDS View Documents
Thermo Scientific Chemicals
437730050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5462-71-5
161.16
WEBXRQONNWEETE-UHFFFAOYSA-N
79587
OC(=O)CC1=CC=C(C=C1)C#N
C9H7NO2
MFCD06798066
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
2-(4-cyanophenyl)acetic acid

Specifications

5462-71-5
100.0
96% min. (HPLC)
C9H7NO2
1g
WEBXRQONNWEETE-UHFFFAOYSA-N
2-(4-cyanophenyl)acetic acid
79587
97%
96.0
Authentic
Glass bottle
MFCD06798066
2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
OC(=O)CC1=CC=C(C=C1)C#N
161.16
161.16
4-Cyanophenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-753-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only