missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanostyrene, 95%, stabilized

CAS: 3435-51-6 | C9H7N | 129.16 g/mol

$308.34 - $308.34

Chemical Identifiers

CAS 3435-51-6
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00080445
InChI Key SNTUCKQYWGHZPK-UHFFFAOYSA-N
PubChem CID 76967
IUPAC Name 4-ethenylbenzonitrile
SMILES C=CC1=CC=C(C=C1)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC292910025
SDS View Documents
Thermo Scientific Chemicals
292910025
2.5 g Glass bottle
Each for $308.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3435-51-6
129.16
SNTUCKQYWGHZPK-UHFFFAOYSA-N
4-ethenylbenzonitrile
C9H7N
MFCD00080445
76967
C=CC1=CC=C(C=C1)C#N

Specifications

3435-51-6
56.0°C to 58.0°C (10.0 mmHg)
94% min. (GC)
C9H7N
MFCD00080445
4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile
SNTUCKQYWGHZPK-UHFFFAOYSA-N
4-ethenylbenzonitrile
0.05% tert.butylcatechol
129.16
Liquid
Colorless to Yellow
Authentic
Glass bottle
1.5765 to 1.5785
2.5 g
Solubility in water: insoluble
C=CC1=CC=C(C=C1)C#N
129.16
76967
95%
4-Cyanostyrene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only