missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Decanol 98.0+%, TCI America™
Supplier: TCI America D138010ML
Specifications
| 4-Decanol | |
| Yellow | |
| C10H22O | |
| 10 mL | |
| DTDMYWXTWWFLGJ-UHFFFAOYSA-N | |
| decan-4-ol | |
| 16320 | |
| ≥98.0% (GC) |
| 2051-31-2 | |
| 211°C | |
| MFCD00039627 | |
| 4-decanol, decanol-4, 4-decanol, s, 4-decyl alcohol, hexylpropylcarbinol, 1-propylheptyl alcohol, acmc-20moz4, acmc-1cf04 | |
| CCCCCCC(CCC)O | |
| 158.285 | |
| 158.29 | |
| Liquid |
Chemical Identifiers
| 2051-31-2 | |
| 158.285 | |
| DTDMYWXTWWFLGJ-UHFFFAOYSA-N | |
| 16320 | |
| CCCCCCC(CCC)O |
| C10H22O | |
| MFCD00039627 | |
| 4-decanol, decanol-4, 4-decanol, s, 4-decyl alcohol, hexylpropylcarbinol, 1-propylheptyl alcohol, acmc-20moz4, acmc-1cf04 | |
| decan-4-ol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No