missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)butyraldehyde Diethyl Acetal 98.0+%, TCI America™
$59.65 - $162.17
Chemical Identifiers
| CAS | 1116-77-4 |
|---|---|
| Molecular Formula | C10H23NO2 |
| Molecular Weight (g/mol) | 189.299 |
| MDL Number | MFCD00671479 |
| InChI Key | QKXMWBLNSPNBEY-UHFFFAOYSA-N |
| Synonym | 4,4-diethoxy-n,n-dimethyl-1-butanamine, 4,4-diethoxy-butyl-dimethyl-amine, 4,4-diethoxy-n,n-dimethylbutylamine, 4,4-diethoxy-n,n-dimethylbutanamine, 4,4-diethoxy-n,n-dimethyl butylamine, 4-n,n-dimethylamino butanal diethyl acetal, 4,4-diethoxy-n, n-dimethyl-butylamine, 4,4-diethoxy-n,n-dimethyl-butan-1-amine, 4-dimethylaminobutyraldehyde diethyl acetal, n,n-dimethyl-4-aminobutanal diethyl acetal |
| PubChem CID | 547078 |
| IUPAC Name | 4,4-diethoxy-N,N-dimethylbutan-1-amine |
| SMILES | CCOC(CCCN(C)C)OCC |
Chemical Identifiers
| 1116-77-4 | |
| 189.299 | |
| QKXMWBLNSPNBEY-UHFFFAOYSA-N | |
| 547078 | |
| CCOC(CCCN(C)C)OCC |
| C10H23NO2 | |
| MFCD00671479 | |
| 4,4-diethoxy-n,n-dimethyl-1-butanamine, 4,4-diethoxy-butyl-dimethyl-amine, 4,4-diethoxy-n,n-dimethylbutylamine, 4,4-diethoxy-n,n-dimethylbutanamine, 4,4-diethoxy-n,n-dimethyl butylamine, 4-n,n-dimethylamino butanal diethyl acetal, 4,4-diethoxy-n, n-dimethyl-butylamine, 4,4-diethoxy-n,n-dimethyl-butan-1-amine, 4-dimethylaminobutyraldehyde diethyl acetal, n,n-dimethyl-4-aminobutanal diethyl acetal | |
| 4,4-diethoxy-N,N-dimethylbutan-1-amine |
Specifications
| 1116-77-4 | |
| 95°C | |
| MFCD00671479 | |
| 4,4-diethoxy-n,n-dimethyl-1-butanamine, 4,4-diethoxy-butyl-dimethyl-amine, 4,4-diethoxy-n,n-dimethylbutylamine, 4,4-diethoxy-n,n-dimethylbutanamine, 4,4-diethoxy-n,n-dimethyl butylamine, 4-n,n-dimethylamino butanal diethyl acetal, 4,4-diethoxy-n, n-dimethyl-butylamine, 4,4-diethoxy-n,n-dimethyl-butan-1-amine, 4-dimethylaminobutyraldehyde diethyl acetal, n,n-dimethyl-4-aminobutanal diethyl acetal | |
| CCOC(CCCN(C)C)OCC | |
| 189.299 | |
| 189.30 | |
| Liquid |
| Yellow | |
| C10H23NO2 | |
| 5 g | |
| QKXMWBLNSPNBEY-UHFFFAOYSA-N | |
| 4,4-diethoxy-N,N-dimethylbutan-1-amine | |
| 547078 | |
| ≥98.0% (GC,T) | |
| 4-(Dimethylamino)butyraldehyde Diethyl Acetal |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator